Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Methoxyphenol, 98+%

Catalog No. p-7023180
Click to view available options
Quantity:
250 g
1000 g
5000 g

2-Methoxyphenol, >-98%, C7H8O2, CAS Number-90-05-1, methylcatechol, guaiastil, 2-hydroxyanisole, 1-hydroxy-2-methoxybenzene, pyroguaiac acid, o-methoxyphenol, pyrocatechol monomethyl ether, phenol, 2-methoxy, guaicol, guaiacol, 1000g, >98%, 508112, 204 deg.C to 206 deg.C, CHEBI:28591, 1.129 | CAS: 90-05-1 | C7H8O2 | 124.139 g/mol

2-Methoxyphenol acts as an expectorant. It is used as an anti-oxidant as well as an anti-skinning agent for paints. It serves as a precursor to flavorants such as eugenol and vanillin. Further, it is used as an intermediate in the chemical synthesis of active pharmaceuticals, flavoring and perfumery products. In addition to this, it is also used as an indicator in chemical reactions that produce oxygen.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Methoxyphenol acts as an expectorant. It is used as an anti-oxidant as well as an anti-skinning agent for paints. It serves as a precursor to flavorants such as eugenol and vanillin. Further, it is used as an intermediate in the chemical synthesis of active pharmaceuticals, flavoring and perfumery products. In addition to this, it is also used as an indicator in chemical reactions that produce oxygen.

Solubility
Miscible with carbon tetrachloride, alcohol, chloroform, ether, oils, glacial acetic acid and sodium hydroxide solution. Slightly miscible with water and petroleum ether.

Notes
Air and light sensitive. Store in a cool place. Keep the container tightly closed in a dry and well-ventilated place. Incompatible with strong oxidizing agents and strong bases.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 90-05-1
Molecular Formula C7H8O2
Molecular Weight (g/mol) 124.139
MDL Number MFCD00002185
InChI Key LHGVFZTZFXWLCP-UHFFFAOYSA-N
Synonym guaiacol, o-methoxyphenol, 2-hydroxyanisole, phenol, 2-methoxy, pyrocatechol monomethyl ether, methylcatechol, guaiastil, guaicol, 1-hydroxy-2-methoxybenzene, pyroguaiac acid
PubChem CID 460
ChEBI CHEBI:28591
IUPAC Name 2-methoxyphenol
SMILES COC1=CC=CC=C1O

Specifications

Melting Point 28°C to 31°C
Density 1.129 g/mL
Boiling Point 204°C to 206°C
Flash Point 82°C (179°F)
Assay Percent Range >98%
Refractive Index 1.543
Quantity 250 g
Beilstein 508112
Sensitivity Air and light sensitive
Merck Index 14,4553
Solubility Information Miscible with carbon tetrachloride,alcohol,chloroform,ether,oils,glacial acetic acid and sodium hydroxide solution. Slightly miscible with water and petroleum ether.
Formula Weight 124.14
Percent Purity ≥98%
Chemical Name or Material 2-Methoxyphenol
Show More Show Less
Hazard Category H227-H302-H315-H319
Hazard Statement GHS H Statement
H302-H315-H319
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
Precautionary Statement P210-P235-P260-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P314-P330-P332+P313-P362-P370+P378q-P501c
EINECSNumber 201-964-7
RTECSNumber SL7525000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.