Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Methoxyphenyl isothiocyanate, 98%

Catalog No. aal1179906
Click to view available options
Quantity:
5 g
25 g

CAS: 3288-04-8 | C8H7NOS | 165.21 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Solubility
Reacts with water.

Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents, amines ansd alcohol. It is sensitive to moisture.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3288-04-8
MDL Number MFCD00011675
ChEBI CHEBI:16318

Specifications

Density 1.196
Boiling Point 267°C
Flash Point >110°C (230°F)
Odor Pungent
Molecular Formula C6H5NO4
Refractive Index 1.6443
Quantity 5 g
UN Number UN2922
Beilstein 638293
Sensitivity Moisture sensitive
Synonym 4-nitrocatechol, 4-nitropyrocatechol, 1,2-dihydroxy-4-nitrobenzene, 4-nitro-1,2-benzenediol, 3,4-dihydroxy-1-nitrobenzene, unii-sw60ng75en, sw60ng75en, 1,2-benzenediol, 4-nitro, nitrocatechol mix of isomers, 4nc
Solubility Information Reacts with water.
InChI Key XJNPNXSISMKQEX-UHFFFAOYSA-N
SMILES C1=CC(=C(C=C1[N+](=O)[O-])O)O
IUPAC Name 4-nitrobenzene-1,2-diol
Molecular Weight (g/mol) 155.11
PubChem CID 3505109
Formula Weight 165.22
Percent Purity 98%
Chemical Name or Material 2-Methoxyphenyl isothiocyanate
Show More Show Less
Hazard Category H301+H311+H331-H315-H319-H335
Hazard Statement GHS H Statement
H331-H314-H318-H302-H312
Toxic if inhaled.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Harmful if swallowed.
Harmful in contact with skin.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P310-P302+P352-P304+P340-P305+P351+P338-P311-P312-P330-P332+P313-P361-P363-P501c
DOTInformation Hazard Class: 6.1; Packaging Group: II
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.