Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Methoxypropene, 97%, stabilized

Catalog No. AC174350010
Click to view available options
Quantity:
250 mL
1 L
2.5 L
Packaging:
Glass bottle

CAS: 116-11-0 | C4H8O | 72.11 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 116-11-0
Molecular Formula C4H8O
Molecular Weight (g/mol) 72.11
MDL Number MFCD00014929
InChI Key YOWQWFMSQCOSBA-UHFFFAOYSA-N
Synonym 2-methoxypropene, isopropenyl methyl ether, 1-propene, 2-methoxy, 2-methoxy-1-propene, methyl isopropenyl ether, ether, isopropenyl methyl, propene, 2-methoxy, unii-15wbg0jt6b, 2-methoxy propene, 15wbg0jt6b
PubChem CID 8300
IUPAC Name 2-methoxyprop-1-ene
SMILES CC(=C)OC

Specifications

CAS Min % 96.0
CAS Max % 100.0
Color Colorless
Density 0.7800g/mL
Boiling Point 35°C to 36°C
Flash Point -29°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Refractive Index 1.3810 to 1.384
Linear Formula CH3C(OCH3)=CH2
Quantity 1 L
Beilstein 01, IV, 2075
Fieser 02,230; 05,360; 11,329; 12,317
Specific Gravity 0.78
Merck Index 15, 6073
Solubility Information Solubility in water: negligible
Stabilizer 0.5% max. potassium carbonate
Formula Weight 72.11
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material 2-Methoxypropene, Stabilized
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Extremely flammable liquid and vapor.
Harmful if swallowed.
May form explosive peroxides.
Precautionary Statement GHS P Statement
Keep container tightly closed.
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Ground/Bond container and receiving equipment.
Take precautionary measures against static discharge.
Wash face
EINECSNumber 204-125-3

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.