Learn More
2-Methyl-1,3-dithiane, 98%
Description
It has been employed in the synthesis of methyl ketones. 2-Methyl-1,3-dithiane is suitable for use in the preparation of potassium trifluoro(2-methyl-1,3-dithiano)borate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
It has been employed in the synthesis of methyl ketones. 2-Methyl-1,3-dithiane is suitable for use in the preparation of potassium trifluoro(2-methyl-1,3-dithiano)borate.
Solubility
Not miscible or difficult to mix in water. Soluble in alcohol.
Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
Specifications
Specifications
| Density | 1.212 |
| Boiling Point | 78°C to 80°C (9 mmHg) |
| Flash Point | 82°C (179°F) |
| Refractive Index | 1.5615 |
| Quantity | 5 g |
| Beilstein | 102766 |
| Solubility Information | Not miscible or difficult to mix in water. Soluble in alcohol. |
| Formula Weight | 134.25 |
| Percent Purity | 98% |
| Chemical Name or Material | 2-Methyl-1,3-dithiane |
Safety and Handling
| Hazard Category | H227 |
| Hazard Statement | GHS H Statement H227 Combustible liquid. |
| Precautionary Statement | P210-P280-P370+P378q-P501c |
| EINECSNumber | 227-859-6 |
| RTECSNumber | JO5263000 |
| TSCA | No |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.