Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Methyl-1-butanethiol, 95%, Thermo Scientific™

Catalog No. p-160645
Click to view available options
Quantity:
25 g
Packaging:
Glass bottle

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1878-18-8
Molecular Formula C5H12S
Molecular Weight (g/mol) 104.21
MDL Number MFCD00004883
InChI Key WGQKBCSACFQGQY-UHFFFAOYNA-N
PubChem CID 15877
IUPAC Name 2-methylbutane-1-thiol
SMILES CCC(C)CS

Specifications

CAS Min % 94.0
CAS Max % 100.0
Color Colorless
Density 0.8400g/mL
Infrared Spectrum Authentic
Assay Percent Range 95%
Packaging Glass bottle
Refractive Index 1.4455 to 1.4475 (20°C, 589nm)
Linear Formula C2H5CH(CH3)CH2SH
Quantity 25 g
Beilstein 01, II, 421
Synonym 2-methyl-1-butanethiol, 1-butanethiol, 2-methyl, 2-methylbutyl mercaptan, 2-methylbutanethiol, fema no. 3303, 2-methyl-butanethiol, acmc-1cuce, 2-methyl-1-butane thiol, 1-butanethiol,2-methyl, 1-mercapto-2-methylbutane
Formula Weight 104.21
Percent Purity ≥94%
Physical Form Liquid
Chemical Name or Material 2-Methyl-1-butanethiol, 95%
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement:
Causes serious eye irritation.
Harmful in contact with skin.
Causes skin irritation.
Harmful if inhaled.
May cause respiratory irritation.
Harmful if swallowed.
Highly flammable liquid and vapor.
Precautionary Statement GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Ground/Bond container and receiving equipment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 217-515-3
TSCA TSCA
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.