Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Methyl-1-buten-3-yne, 98%, ACROS Organics™

Click to view available options
Quantity:
5mL
10mL
25mL
50mL
78-80-8
C5H6
66.1
MFCD00008599
BOFLDKIFLIFLJA-UHFFFAOYSA-N
isopropenylacetylene,2-methyl-1-buten-3-yne,valylene,1-buten-3-yne, 2-methyl,3-methyl-3-buten-1-yne,2-methylbutenyne,isopropenyl acetylene,acmc-20anrd,2-methyl-1-butenyne,2-methyl-l-buten-3-yne
62323
2-methylbut-1-en-3-yne
CC(=C)C#C
78-80-8
C5H6
66.1
MFCD00008599
BOFLDKIFLIFLJA-UHFFFAOYSA-N
isopropenylacetylene,2-methyl-1-buten-3-yne,valylene,1-buten-3-yne, 2-methyl,3-methyl-3-buten-1-yne,2-methylbutenyne,isopropenyl acetylene,acmc-20anrd,2-methyl-1-butenyne,2-methyl-l-buten-3-yne
62323
2-methylbut-1-en-3-yne
CC(=C)C#C
Melting Point -113°C
Boiling Point 32°C
Flash Point -45°C
Specific Gravity 0.69
Density 0.6900g/mL
Refractive Index 1.4140 to 1.416
Color Undesignated
Packaging Glass bottle
Quantity 50mL
Infrared Spectrum Authentic
CAS Min % 97.5
CAS Max % 100.0
Assay Percent Range 97.5% min. (GC)
Solubility Information Solubility in water: .. Other solubilities: soluble in organic solvents
Formula Weight 66.1
Physical Form Liquid
Percent Purity 98%
Chemical Name or Material 2-Methyl-1-buten-3-yne
Show More Show Less

GHS H Statement:
Causes serious eye irritation.
Harmful if inhaled.
Harmful if swallowed.
Causes skin irritation.
May cause respiratory irritation.
Extremely flammable liquid and vapor.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Ground/Bond container and receiving equipment.
Take precautionary measures against static discharge.
Store in a well-ventilated place. Keep cool.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger

EINECSNumber : 201-144-9