Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Methyl-1-morpholino-2-propanamine, 97%, Thermo Scientific™

Catalog No. MO01187CB
Change view
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Amber Glass Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
MO01187CB 250 mg Amber Glass Bottle
MO01187DA 1 g Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. MO01187CB Supplier Thermo Scientific Chemicals Supplier No. MO01187CB
Add to Cart

Chemical Identifiers

CAS 6105-75-5
Molecular Formula C8H18N2O
Molecular Weight (g/mol) 158.25
MDL Number MFCD01714129
InChI Key JCXYZQUYVNLCTG-UHFFFAOYSA-N
Synonym 2-methyl-1-morpholinopropan-2-amine, 2-methyl-1-morpholin-4-yl propan-2-amine, 1,1-dimethyl-2-morpholin-4-yl-ethylamine, 2-methyl-1-morpholino-2-propanamine, 4-2-amino-2-methylpropyl morpholine, morpholine, 4-2-amino-2-methylpropyl, morpholine, n-2-aminoisobutyl, 1,1-dimethyl-2-morpholin-4-ylethylamine, 1,1-dimethyl-2-morpholin-4-ylethyl amine, 4-morpholineethanamine, alpha,alpha-dimethyl
PubChem CID 201497
IUPAC Name 2-methyl-1-morpholin-4-ylpropan-2-amine
SMILES CC(C)(N)CN1CCOCC1

Specifications

CAS Min % 97.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 250 mg
Formula Weight 158.24
Percent Purity 97%
Chemical Name or Material 2-Methyl-1-morpholino-2-propanamine
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement:
Causes severe skin burns and eye damage.
Causes serious eye damage.
Precautionary Statement GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.