Learn More
2-Methyl-1-pentene, 97%
Description
2-Methyl-1-pentene is prepared from 2-Methyl-1-pentene. 2-methyl-1-pentene is used in copolymerization of ethylene catalyzed by nonbridged half-titanocenes. The isomerization of 2-methyl-1-pentene leads to anionic activation of c-h bonds in olefins.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
2-Methyl-1-pentene is prepared from 2-Methyl-1-pentene. 2-methyl-1-pentene is used in copolymerization of ethylene catalyzed by nonbridged half-titanocenes. The isomerization of 2-methyl-1-pentene leads to anionic activation of c-h bonds in olefins.
Solubility
Insoluble in water.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
Specifications
Specifications
| Melting Point | -136°C |
| Density | 0.682 |
| Boiling Point | 62°C |
| Flash Point | −27°C (−17°F) |
| Refractive Index | 1.392 |
| Quantity | 25 mL |
| UN Number | UN2288 |
| Beilstein | 1340499 |
| Solubility Information | Insoluble in water. |
| Formula Weight | 84.16 |
| Show More |
Safety and Handling
| Hazard Category | H225-H304-H336 |
| Hazard Statement | GHS H Statement H225 Highly flammable liquid and vapor. |
| Precautionary Statement | P210-P233-P235-P240-P241-P242-P243-P261-P271-P280-P301+P310-P303+P361+P353-P304+P340-P312-P331-P370+P378q-P501c |
| DOTInformation | Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: ISOHEXENES |
| EINECSNumber | 212-108-7 |
| RTECSNumber | SB2230000 |
| TSCA | Yes |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.