Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Methyl-1-pentene, 99%

Catalog No. p-166351
Click to view available options
Quantity:
10 mL
50 mL
Packaging:
Glass bottle

CAS: 763-29-1 | C6H12 | 84.15 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 763-29-1
Molecular Formula C6H12
Molecular Weight (g/mol) 84.15
MDL Number MFCD00009405
InChI Key WWUVJRULCWHUSA-UHFFFAOYSA-N
Synonym 2-methyl-1-pentene, 2-methylpentene, 1-pentene, 2-methyl, 4-methyl-4-pentene, 2-methyl-pentene-1, unii-okc25o38mc, okc25o38mc, acmc-1bixb, 4-01-00-00841 beilstein handbook reference, c2h5ch2c ch3 =ch2
PubChem CID 12986
IUPAC Name 2-methylpent-1-ene
SMILES CCCC(=C)C

Specifications

CAS Min % 98.5
CAS Max % 100.0
Melting Point -136.0°C
Color Colorless
Density 0.6800g/mL
Boiling Point 62.0°C
Flash Point −26°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Linear Formula CH3CH2CH2C(CH3)=CH2
Refractive Index 1.3910 to 1.9300
Quantity 10 mL
Beilstein 01, I, 90
Specific Gravity 0.68
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in alcohol, acetone and ether
Formula Weight 84.15
Percent Purity 99%
Physical Form Liquid
Chemical Name or Material 2-Methyl-1-pentene, 99%
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Highly flammable liquid and vapor.
May be fatal if swallowed and enters airways.
May cause drowsiness or dizziness.
Precautionary Statement GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
IF ON SKIN (or hair): Take off immediately all contaminated clothing.
Rinse skin with water/shower.
IF SWALLOWED: Immediately call a POISON CENTER o
EINECSNumber 212-108-7
RTECSNumber SB2230000
TSCA TSCA

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.