Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Methyl-1-propanol, 99+%, Extra Dry, AcroSeal™

Catalog No. p-3759070
Click to view available options
Quantity:
100 mL
1 L
This item is not returnable. View return policy
This item is not returnable. View return policy

CAS: 78-83-1 | C4H10O | 74.12 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 78-83-1
Molecular Formula C4H10O
Molecular Weight (g/mol) 74.12
MDL Number MFCD00004740
InChI Key ZXEKIIBDNHEJCQ-UHFFFAOYSA-N
Synonym 2-methyl-1-propanol, isobutanol, isobutyl alcohol, 1-propanol, 2-methyl, 1-hydroxymethylpropane, isopropylcarbinol, iso-butyl alcohol, 2-methylpropyl alcohol, isobutylalkohol, i-butyl alcohol
PubChem CID 6560
ChEBI CHEBI:46645
IUPAC Name 2-methylpropan-1-ol
SMILES CC(C)CO

Specifications

CAS Min % 99.0
CAS Max % 100.0
Melting Point -108.0°C
Density 0.8000g/mL
Boiling Point 108.0°C
Flash Point 28°C
Assay Percent Range 99% min. (GC)
Packaging Glass Bottle
Refractive Index 1.3945 to 1.3965
Linear Formula (CH3)2CHCH2OH
Quantity 100 mL
Beilstein 01, 373
Specific Gravity 0.8
Merck Index 15, 5176
Solubility Information Solubility in water: 95g/L (20°C). Other solubilities: soluble in alcohol, ether, acetone
Viscosity 4 Pas (20°C)
Formula Weight 74.12
Percent Purity 99+%
Grade Extra Dry
Chemical Name or Material 2-Methyl-1-propanol, Extra Dry
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Causes serious eye damage.
May cause respiratory irritation.
May cause drowsiness or dizziness.
Causes skin irritation.
Flammable liquid and vapor.
Precautionary Statement GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
IF ON SKIN (or hair): Take off immediately all contaminated clothing.
Rinse skin with water/shower.
If skin irritation occurs: Get medical advice/at
EINECSNumber 201-148-

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.