Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Methyl-1-propanol, Analytical Standard, ≥99.8% (GC), MilliporeSigma™ Supelco™

Catalog No. 111018790
Click to view available options
Quantity:
1 mL
5 mL
10 mL

2-Methyl-1-propanol can be obtained via hydroformylation of alkenes with fuel gas mixture in particular, carbon monoxide being the main reactant.

TRUSTED_SUSTAINABILITY

Specifications

CAS 78-83-1
Vapor Pressure 8.8 mmHg (0°C)
Vapor Density 2.55 (vs. air)
Density 0.803 g/mL (at 25°C)
Physical Form Neat
Linear Formula (CH3)2 CHCH2OH
MDL Number MFCD00004740
Melting Point -108°C
Boiling Point 108°C
Synonym Isobutanol; Isobutyl alcohol
Quantity 5 mL
Formula Weight 74.12
Percent Purity ≥99.8% (GC)
Grade Analytical Standard
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.