Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Methyl-2,3,4,5-tetrahydro-1H-pyrido[4,3-b]indole 98.0+%, TCI America™
SDP

Catalog No. M27171G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
200 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M27171G 1 g
M2717200MG 200 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M27171G Supplier TCI America Supplier No. M27171G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 5094-12-2
Molecular Formula C12H14N2
Molecular Weight (g/mol) 186.26
MDL Number MFCD00460783
InChI Key FYHWPFXPFFPQRT-UHFFFAOYSA-N
PubChem CID 97424
IUPAC Name 2-methyl-1,3,4,5-tetrahydropyrido[4,3-b]indole
SMILES CN1CCC2=C(C1)C3=CC=CC=C3N2

Specifications

Melting Point 172°C
Color White-Yellow
Quantity 1 g
Formula Weight 186.26
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 2-Methyl-2,3,4,5-tetrahydro-1H-pyrido[4,3-b]indole
RTECSNumber UV0830000
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.