Learn More
(+/-)-2-Methyl-2,4-pentanediol, 98%
Description
(±)-2-Methyl-2,4-pentanediol is used to control the flow properties of industrial products such as paints, coatings, cleansers, solvents, and hydraulic fluids. It acts as a thickening agent in cosmetic products. It serves as a coupling agent and an additive to hydraulic fluids, inks and cement. It is also employed as a blood volume expander. Further, it is used as a solvent for cleaning and colorant products. In addition to this, it is used in laboratory studies as a precipitant and cryoprotectant in protein crystallography.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
(^+)-2-Methyl-2,4-pentanediol is used to control the flow properties of industrial products such as paints, coatings, cleansers, solvents, and hydraulic fluids. It acts as a thickening agent in cosmetic products. It serves as a coupling agent and an additive to hydraulic fluids, inks and cement. It is also employed as a blood volume expander. Further, it is used as a solvent for cleaning and colorant products. In addition to this, it is used in laboratory studies as a precipitant and cryoprotectant in protein crystallography.
Solubility
Miscible with water, alcohol, ether, fatty acids and lower aliphatic hydrocarbons. Slightly miscible with carbon tetrachloride.
Notes
Hygroscopic. Incompatible with strong acids, strong oxidizing agents and strong reducing agents.
Specifications
Specifications
| Melting Point | -40°C |
| Density | 0.922 |
| Boiling Point | 197°C |
| Flash Point | 93°C (199°F) |
| Odor | Weak |
| Refractive Index | 1.427 |
| Quantity | 2500 mL |
| Beilstein | 1098298 |
| Sensitivity | Hygroscopic |
| Merck Index | 14,4710 |
| Show More |
Safety and Handling
| Hazard Category | H227-H315-H319-X |
| Hazard Statement | GHS H Statement H315-H319-H227 Causes skin irritation. Causes serious eye irritation. Combustible liquid. |
| Precautionary Statement | P210-P233-P240-P243-P260-P264b-P271-P280-P285-P301+P310-P302+P352-P304+P340-P305+P351+P338-P332+P313-P362-P374-P501c |
| EINECSNumber | 203-489-0 |
| RTECSNumber | SA0810000 |
| TSCA | Yes |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.