Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Methyl-2,4-pentanediol, 99%, ACROS Organics™

Click to view available options
Quantity:
1L
2.5L
25L
Packaging:
Glass Bottle
Metal drum
107-41-5
C6H14O2
118.18
MFCD00004547
SVTBMSDMJJWYQN-UHFFFAOYSA-N
hexylene glycol,2-methyl-2,4-pentanediol,pinakon,2,4-dihydroxy-2-methylpentane,2,4-pentanediol, 2-methyl,diolane,isol,4-methyl-2,4-pentanediol,1,1,3-trimethyltrimethylenediol,2-methyl pentane-2,4-diol
7870
CHEBI:62995
2-methylpentane-2,4-diol
CC(CC(C)(C)O)O
107-41-5
C6H14O2
118.18
MFCD00004547
SVTBMSDMJJWYQN-UHFFFAOYSA-N
hexylene glycol,2-methyl-2,4-pentanediol,pinakon,2,4-dihydroxy-2-methylpentane,2,4-pentanediol, 2-methyl,diolane,isol,4-methyl-2,4-pentanediol,1,1,3-trimethyltrimethylenediol,2-methyl pentane-2,4-diol
7870
CHEBI:62995
2-methylpentane-2,4-diol
CC(CC(C)(C)O)O
Viscosity 36 mPa.s (20°C)
Melting Point -40.0°C
Quantity 1L
Packaging Glass Bottle
Boiling Point 197.0°C
Density 0.9220g/mL
Color Undesignated
Flash Point 93°C
Specific Gravity 0.922
Refractive Index 1.4260 to 1.4280
Infrared Spectrum Authentic
CAS Min % 98.5
CAS Max % 100.0
Assay Percent Range 99%
Linear Formula CH3CH(OH)CH2C(CH3)2OH
Beilstein 01, 486
Fieser 01,693
Merck Index 15, 4746
Solubility Information Solubility in water: soluble. Other solubilities: soluble in alcohol,ether and lower aliphathic,hydrocarbons
Formula Weight 118.18
Physical Form Slightly Viscous Liquid
Percent Purity 99%
Chemical Name or Material 2-Methyl-2, 4-pentanediol
Show More Show Less

GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.

GHS P Statement:
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.