Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Methyl-2-butanol, ≥99.5% (GC), MilliporeSigma™ Supelco™

Catalog No. p-7187999
Click to view available options
Quantity:
1 mL
5 mL

2-Methyl-2-butanol is an isomer of n-pentanol. It is an organic solvent used mostly in esterification.

Featured Industries: Cleaning Products, Cosmetics, Environmental, Food and Beverages and Personal Care
TRUSTED_SUSTAINABILITY

Specifications

Ignition Point 819°F
CAS 75-85-4
Vapor Density 3 (vs. air)
Density 0.805 g/mL (at 25°C)
Flash Point 20.5°C - Closed Cup
Physical Form Neat
Linear Formula CH3 CH2 C(CH3)2 OH
MDL Number MFCD00004478
Melting Point -12°C
Boiling Point 102°C
Synonym tert-Amyl alcohol; tert-Pentyl alcohol
Type Analytical Standard
Quantity 1 mL
Formula Weight 88.15
Percent Purity ≥99.5% (GC)
Grade Analytical Standard
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.