Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Methyl-2-butene, ≥99% (GC), MilliporeSigma™ Supelco™

Catalog No. 111019097 Shop All MilliporeSigma Supelco Products
Click to view available options
Quantity:
5 mL
10 mL
50 mL

2-Methyl-2-butene is a trisubstituted olefin which is mostly used as a precursor for peroxyacetyl nitrate. It can be prepared by the acid-catalyzed dehydration of 3-methyl-2-butanol. It has 3 alkyl groups on the double bond.

Featured Industries: Environmental and Petroleum
TRUSTED_SUSTAINABILITY

Specifications

CAS 513-35-9
Vapor Density 2.4 (vs. air)
Density 0.662 g/mL (at 25°C)
Flash Point -20°C
Physical Form Neat
Linear Formula CH3 CH=C(CH3)2
MDL Number MFCD00009276
Melting Point -134°C
Boiling Point 35°C to 38°C
Synonym beta-Isoamylene; Amylene
Type Analytical Standard
Quantity 10 mL
Formula Weight 70.13
Percent Purity ≥99% (GC)
Grade Analytical Standard
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.