Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Methyl-2-oxazoline, 99%

Catalog No. p-166382
Click to view available options
Quantity:
100 g
500 g
Packaging:
Glass bottle

CAS: 1120-64-5 | C4H7NO | 85.11 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1120-64-5
Molecular Formula C4H7NO
Molecular Weight (g/mol) 85.11
MDL Number MFCD00005298
InChI Key GUXJXWKCUUWCLX-UHFFFAOYSA-N
Synonym 2-methyl-2-oxazoline, 2-methyloxazoline, 2-methyl-4,5-dihydrooxazole, oxazole, 4,5-dihydro-2-methyl, 2-methyl-2-oxazolin, 2-methyl-1,3-oxazoline, 2-oxazoline, 2-methyl, 4,5-dihydro-2-methyl-1,3-oxazole, poly 2-methyl-2-oxazoline, methyl oxazoline
PubChem CID 70713
ChEBI CHEBI:53614
IUPAC Name 2-methyl-4,5-dihydro-1,3-oxazole
SMILES CC1=NCCO1

Specifications

CAS Min % 98.5
CAS Max % 100.0
Color Colorless to Yellow
Density 1.0000g/mL
Boiling Point 109.4°C to 110.4°C
Flash Point 20°C
Infrared Spectrum Authentic
Assay Percent Range 99%
Packaging Glass bottle
Refractive Index 1.4330 to 1.4350
Quantity 100 g
Beilstein 27, 13
Specific Gravity 1
Formula Weight 85.11
Percent Purity 99%
Physical Form Liquid
Chemical Name or Material 2-Methyl-2-oxazoline, 99%
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement:
Highly flammable liquid and vapor.
Precautionary Statement GHS P Statement:
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Ground/Bond container and receiving equipment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 214-316-3
TSCA TSCA

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.