Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Methyl-2-pentenal, 97%, ACROS Organics™

Click to view available options
Quantity:
25g
100g
Packaging:
Glass bottle
623-36-9
C6H10O
98.15
MFCD00006978
IDEYZABHVQLHAF-GQCTYLIASA-N
2-methyl-2-pentenal,2-pentenal, 2-methyl,e-2-methyl-2-pentenal,2-methyl-3-ethylacrolein,2-methyl-2-penten-1-al,2-methyl-2-pentene-1-al,2,4-dimethylcrotonaldehyde,trans-2-methyl-2-pentenal,e-2-methylpent-2-enal,unii-98qag1u530
5319754
(E)-2-methylpent-2-enal
CCC=C(C)C=O
623-36-9
C6H10O
98.15
MFCD00006978
IDEYZABHVQLHAF-GQCTYLIASA-N
2-methyl-2-pentenal,2-pentenal, 2-methyl,e-2-methyl-2-pentenal,2-methyl-3-ethylacrolein,2-methyl-2-penten-1-al,2-methyl-2-pentene-1-al,2,4-dimethylcrotonaldehyde,trans-2-methyl-2-pentenal,e-2-methylpent-2-enal,unii-98qag1u530
5319754
(E)-2-methylpent-2-enal
CCC=C(C)C=O
Color Colorless to Yellow
Density 0.8600g/mL
Packaging Glass bottle
Infrared Spectrum Authentic
Refractive Index 1.4470 to 1.4550
Boiling Point 134.0°C to 136.0°C
Specific Gravity 0.86
Quantity 100g
Flash Point 31°C
CAS Min % 96.0
CAS Max % 100.0
Assay Percent Range 96% min. (GC)
Linear Formula C2H5CH=C(CH3)CHO
Solubility Information Solubility in water: insoluble
Formula Weight 98.15
Physical Form Liquid
Percent Purity 97%
Chemical Name or Material 2-Methyl-2-pentenal
Show More Show Less

GHS H Statement
Toxic if inhaled.
May cause an allergic skin reaction.
Causes serious eye damage.
Flammable liquid and vapor.

GHS P Statement
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.

GHS Signal Word: Danger

EINECSNumber : 210-789-5