Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Methyl-3,5-dinitrobenzoic acid, 97+%

Catalog No. aaa1455514
Click to view available options
Quantity:
25 g
100 g
500 g

CAS: 28169-46-2 | C8H6N2O6 | 226.14 g/mol

2-Methyl-3,5-dinitrobenzoic acid was used to synthesis heterocyclic derivatives and in the synthesis of isoquinolone.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Methyl-3,5-dinitrobenzoic acid was used to synthesis heterocyclic derivatives and in the synthesis of isoquinolone.

Solubility
Soluble in water.

Notes
Store in cool, dry conditions in a well sealed container. Store away from oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 28169-46-2
Molecular Formula C8H6N2O6
Molecular Weight (g/mol) 226.14
MDL Number MFCD00007161
InChI Key CDVNZMKTJIBBBV-UHFFFAOYSA-N
Synonym 2-methyl-3,5-dinitrobenzoic acid, 3,5-dinitro-2-methylbenzoic acid, 3,5-dinitro-o-toluic acid, benzoic acid, 2-methyl-3,5-dinitro, 3,5-dinitrotoluic acid, o-toluic acid, 3,5-dinitro, 2-methyl-3,5-dinitro-benzoic acid, 3,5-dinitro-2-methyl-benzoic acid, acmc-1cfeu, o-toluic acid,5-dinitro
PubChem CID 98824
IUPAC Name 2-methyl-3,5-dinitrobenzoic acid
SMILES CC1=C(C=C(C=C1[N+]([O-])=O)[N+]([O-])=O)C(O)=O

Specifications

Melting Point 203°C to 207°C
Quantity 25 g
Beilstein 2622906
Solubility Information Soluble in water.
Formula Weight 226.15
Percent Purity 98%
Chemical Name or Material 2-Methyl-3,5-dinitrobenzoic acid
Hazard Category H302-H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
EINECSNumber 248-880-7
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.