Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Methyl-3-(methylthio)pyrazine 99.0+%, TCI America™
SDP

Catalog No. m06851ml Shop All Tokyo Chemical Industry (TCI) Products
Click to view available options
Quantity:
1 mL

Chemical Identifiers

CAS 2882-20-4
Molecular Formula C6H8N2S
Molecular Weight (g/mol) 140.20
MDL Number MFCD00014610
InChI Key PPPFFGVGWFKTHX-UHFFFAOYSA-N
Synonym 2-methyl-3-methylthio pyrazine, 2-methylthio-3-methylpyrazine, 2-methyl-3-methylsulfanyl-pyrazine, pyrazine, 2-methyl-3-methylthio, 2-methylmercapto-3-methylpyrazine, methylthio methyl pyrazine, 3-methyl-2-methylthiopyrazine, 2-methyl-3-methylmercaptopyrazine, unii-ki350cs0lz, pyrazine, 2-methyl-3 or 5-methylthio
PubChem CID 76152
IUPAC Name 2-methyl-3-(methylsulfanyl)pyrazine
SMILES CSC1=NC=CN=C1C

Specifications

Color Colorless
Boiling Point 104°C
Quantity 1 mL
Formula Weight 140.20
Percent Purity ≥99.0% (GC)
Physical Form Liquid
Chemical Name or Material 2-Methyl-3-(methylthio)pyrazine
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.