Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Aminoacetaldehyde dimethyl acetal, 99%

Catalog No. AC153341000
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
2.5 kg
Packaging:
Glass bottle
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC153341000 100 g Glass bottle
AC153340250 25 g Glass bottle
AC153345000 500 g Glass bottle
AC153340025 2.5 kg Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Catalog No. AC153341000 Supplier Thermo Scientific Chemicals Supplier No. 153341000
Only null left
Add to Cart
This item is not returnable. View return policy

CAS: 22483-09-6 | C4H11NO2 | 105.14 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 22483-09-6
MDL Number MFCD00008135
InChI Key QKWWDTYDYOFRJL-UHFFFAOYSA-N
Synonym aminoacetaldehyde dimethyl acetal, 2,2-dimethoxyethylamine, ethanamine, 2,2-dimethoxy, 2,2-dimethoxyethan-1-amine, dimethylaminoacetal, amino acetaldehyde dimethyl acetal, 1-amino-2,2-dimethoxyethane, 2-aminoacetaldehyde dimethyl acetal, acetaldehyde, amino-, dimethyl acetal, acmc-209fwr
PubChem CID 89728
IUPAC Name 2,2-dimethoxyethanamine
SMILES COC(CN)OC

Specifications

Color Colorless to Yellow
Density 0.9700g/mL
Boiling Point 135°C to 139°C (95.0 mmHg)
Flash Point 41°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Molecular Formula C4H11NO2
Refractive Index 1.4160 to 1.418
Linear Formula H2NCH2CH(OCH3)2
Quantity 100 g
Beilstein 04, II, 758
Specific Gravity 0.97
Solubility Information Solubility in water: miscible
Molecular Weight (g/mol) 105.14
Viscosity 0.15 Pas (20°C)
Formula Weight 105.14
Percent Purity 99%
Physical Form Liquid
Chemical Name or Material Aminoacetaldehyde dimethyl acetal, 99%
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Flammable liquid and vapor.
Causes severe skin burns and eye damage.
Precautionary Statement GHS P Statement
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsi
EINECSNumber 245-026-5
TSCA TSCA

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.