Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Methyl-3-tetrahydrothiophenone 97.0+%, TCI America™
SDP

Catalog No. M183925G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M183925G 25 g
M18395G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M183925G Supplier TCI America Supplier No. M183925G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 13679-85-1
Molecular Formula C5H8OS
Molecular Weight (g/mol) 116.178
MDL Number MFCD00078280
InChI Key YMZZPMVKABUEBL-UHFFFAOYSA-N
Synonym 2-methyltetrahydrothiophen-3-one, dihydro-2-methyl-3 2h-thiophenone, 2-methyldihydrothiophen-3 2h-one, 3 2h-thiophenone, dihydro-2-methyl, 2-methyltetrahydrothiophene-3-one, 2-methyl-3-oxotetrahydrothiophene, 2-methyl-4,5-dihydro-3 2h-thiophenone, 2-methyl-3-tetrahydrothiophenone, 2-methyl tetrahydrothiophen-3-one, xi-dihydro-2-methyl-3 2h-thiophenone
PubChem CID 61664
ChEBI CHEBI:87506
IUPAC Name 2-methylthiolan-3-one
SMILES CC1C(=O)CCS1

Specifications

Color Yellow
Boiling Point 86°C
Quantity 25 g
Formula Weight 116.18
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material 2-Methyl-3-tetrahydrothiophenone
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.