Learn More
2-Methyl-4-phenyl-5-pyrimidinecarboxylic acid, 97%, Thermo Scientific™
Specifications
Specifications
| CAS Min % | 96.5 |
| CAS Max % | 100.0 |
| Assay Percent Range | 97% |
| Packaging | Amber glass bottle |
| Quantity | 1 g |
| Synonym | 2-methyl-4-phenyl-5-pyrimidinecarboxylic acid, 5-pyrimidinecarboxylicacid, 2-methyl-4-phenyl, 5-pyrimidinecarboxylic acid, 2-methyl-4-phenyl, acmc-1c0qx, 2-methyl-4-phenylpyrimidin-5-carboxylic acid, 5-pyrimidinecarboxylicacid,2-methyl-4-phenyl, 2-methyl-4-phenyl-pyrimidine-5-carboxylic acid |
| Formula Weight | 214.22 |
| Percent Purity | 97% |
| Chemical Name or Material | 2-Methyl-4-phenyl-5-pyrimidinecarboxylic acid |
Safety and Handling
| Hazard Statement | GHS H Statement: Causes skin irritation. Causes serious eye irritation. May cause respiratory irritation. |
| Precautionary Statement | GHS P Statement: Avoid breathing dust/fume/gas/mist/vapors/spray. IF ON SKIN: Wash with plenty of soap and water. Wear protective gloves/protective clothing/eye protection/face protection. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification. |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.