Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Methyl-5-hydroxytryptamine hydrochloride, Tocris Bioscience™

Catalog No. p-4783168
Click to view available options
Quantity:
10 mg
50 mg
This item is not returnable. View return policy
This item is not returnable. View return policy

5-HT3 agonist/potent 5-HT6 ligand

5-HT3 agonist (Ki = 1200 nM) and potent 5-HT6 ligand (Ki = 46 nM).

Chemical Identifiers

CAS 845861-49-6
Molecular Formula C11H15ClN2O
Molecular Weight (g/mol) 226.704
InChI Key ZNVRHQPXSBWLQP-UHFFFAOYSA-N
Synonym 2-methylserotonin.hcl, 2-methyl-5-hydroxytryptamine hydrochloride, 3-2-aminoethyl-2-methyl-1h-indol-5-ol hydrochloride, 2-methyl-5-ht hydrochloride
PubChem CID 11492320
IUPAC Name 3-(2-aminoethyl)-2-methyl-1H-indol-5-ol;hydrochloride
SMILES CC1=C(C2=C(N1)C=CC(=C2)O)CCN.Cl

Specifications

Quantity 10 mg
Formula Weight 226.71
Percent Purity >98%
Chemical Name or Material 2-Methyl-5-hydroxytryptamine hydrochloride
Recommended Storage Store at Room Temperature
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.