Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
L-Alanine, 99%, specified according to the requirements of Ph.Eur.
Click to view available options
Quantity:
250 g
1 kg
2.5 kg
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications
Specifications
Melting Point | 314.0°C to 316.0°C |
Assay Percent Range | 99% |
Linear Formula | CH3CH(NH2)CO2H |
Specific Rotation | + 14.50 |
Solubility Information | Solubility in water: 166.5g/l in water (25°C). Other solubilities: slightly soluble in alcohol, insoluble in ether and organic solvents |
Quantity | 1 kg |
Formula Weight | 89.09 |
Percent Purity | 99% |
Chemical Name or Material | L-Alanine, specified according to the requirements of Ph.Eur. |
RUO – Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction