Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Methyl-7-[phenyl(phenylamino)methyl]-8-quinolinol 98.0+%, TCI America™
SDP

Catalog No. M2390100MG Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 mg
100 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M2390100MG 100 mg
M239025MG 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M2390100MG Supplier TCI America Supplier No. M2390100MG
Add to Cart
Add to Cart

Chemical Identifiers

CAS 6964-62-1
Molecular Formula C23H20N2O
Molecular Weight (g/mol) 340.426
MDL Number MFCD08823776
InChI Key WEENRMPCSWFMTE-UHFFFAOYSA-N
Synonym 2-methyl-7-phenyl phenylamino methyl-8-quinolinol, 7-anilino phenyl methyl-2-methylquinolin-8-ol, 7-anilino phenyl-methyl-2-methyl-8-quinolinol, 2-methyl-7-phenyl phenylamino methyl quinolin-8-ol, 8-hydroxy-2-methyl-7-phenyl phenylamino methyl quinoline, d0c0tn, 7-anilino phenyl methyl-2-methyl-8-quinolinol, 2-methyl-7-phenyl-phenylamino methyl quinolin-8-ol, 8-quinolinol,2-methyl-7-phenyl phenylamino methyl
PubChem CID 248986
IUPAC Name 7-[anilino(phenyl)methyl]-2-methylquinolin-8-ol
SMILES CC1=NC2=C(C=C1)C=CC(=C2O)C(C3=CC=CC=C3)NC4=CC=CC=C4

Specifications

Melting Point 145°C
Quantity 100 mg
Formula Weight 340.43
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2-Methyl-7-[phenyl(phenylamino)methyl]-8-quinolinol
TSCA No
Recommended Storage Refrigerator
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.