Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 2-(Methylamino)ethanol, 99%

Manufacturer:  Alfa Aesar™ B23180AE

Quantity 100mL

 View more versions of this product

Catalog No. AAB23180AE


  Check Availability
Add to cart

Specifications

Description

Applications
2-(Methylamino)ethanol is used as an intermediate in synthetic chemistry. It finds application in various fields such as textile lubricants, polishes, detergents and in personal care products. It is also involved in electrostatic automotive coatings and in acid gas scrubbing. Further, it serves as an intermediate in polymers and pharmaceuticals. In addition to this, it is used as a solvent in the natural gas-processing industry.

Solubility
Miscible with water, alcohol and ether.

Notes
Incompatible with oxidizing agents, acids, copper, copper alloys, zinc and galvanized iron.
Description & Specifications

Chemical Identifiers

109-83-1
CH3NH(CH2)2OH
MFCD00002839
1071196
N-Methylethanolamine
C5H7NO
75.11
UN2735
14,6015
2-(Methylamino)ethanol

Specifications

109-83-1
CH3NH(CH2)2OH
MFCD00002839
1071196
N-Methylethanolamine
159°C
74°C (165°F)
2-(Methylamino)ethanol
C5H7NO
75.11
UN2735
14,6015
−5°C
100mL
Ammonia-like
99%

Safety and Handling

SDS & Safety Documents

GHS H Statement
H314-H302-H312-H227
Causes severe skin burns and eye damage.
Harmful if swallowed.
Harmful in contact with skin.
Combustible liquid.

GHS P Statement
P210-P260-P303+P361+P353-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: AMINES, LIQUID, CORROSIVE, N.O.S.

RTECSNumber : KL6650000

TSCA : TSCA

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit