Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-(Methylamino)pyridine, 98%

Catalog No. AAB2358914
Click to view available options
Quantity:
1 g
5 g
25 g

CAS: 4597-87-9 | C6H8N2 | 108.14 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 4597-87-9
Molecular Formula C6H8N2
Molecular Weight (g/mol) 108.14
MDL Number MFCD00006250
InChI Key SVEUVITYHIHZQE-UHFFFAOYSA-N
Synonym 2-methylamino pyridine, 2-methylaminopyridine, 2-pyridinamine, n-methyl, methylamino-2 pyridine, n-2-pyridylmethylamine, pyridine, 2-methylamino, n-methyl-2-pyridinamine, 2-pyridinamine, n-methyl-9ci, n-methyl pyridimine
PubChem CID 20727
IUPAC Name N-methylpyridin-2-amine
SMILES CNC1=CC=CC=N1

Specifications

Melting Point 14°C to 15°C
Density 1.06
Boiling Point 200°C to 201°C
Flash Point 87°C (188°F)
Refractive Index 1.578
Quantity 25 g
UN Number UN2810
Beilstein 108188
Formula Weight 108.14
Percent Purity 98%
Chemical Name or Material 2-(Methylamino)pyridine
Show More Show Less
Hazard Category H227-H302+H312+H332-H315-H319-H335
Hazard Statement GHS H Statement
H331-H227-H302-H312-H315-H319-H335
Toxic if inhaled.
Combustible liquid.
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P210-P235-P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P370+P378q-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC, LIQUIDS, ORGANIC, N.O.S.
EINECSNumber 224-997-9
RTECSNumber UT5500000
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.