Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Methylanisole, 99%, ACROS Organics™

Click to view available options
Quantity:
5g
100g
500g
Packaging:
Glass bottle
578-58-5
C8H10O
122.17
MFCD00008373
DTFKRVXLBCAIOZ-UHFFFAOYSA-N
2-methylanisole,o-methylanisole,2-methoxytoluene,o-cresol methyl ether,o-cresyl methyl ether,anisole, o-methyl,benzene, 1-methoxy-2-methyl,methyl o-tolyl ether,o-methoxytoluene,2-methylmethoxybenzene
33637
1-methoxy-2-methylbenzene
CC1=CC=CC=C1OC
578-58-5
C8H10O
122.17
MFCD00008373
DTFKRVXLBCAIOZ-UHFFFAOYSA-N
2-methylanisole,o-methylanisole,2-methoxytoluene,o-cresol methyl ether,o-cresyl methyl ether,anisole, o-methyl,benzene, 1-methoxy-2-methyl,methyl o-tolyl ether,o-methoxytoluene,2-methylmethoxybenzene
33637
1-methoxy-2-methylbenzene
CC1=CC=CC=C1OC
Refractive Index 1.5160 to 1.518
Flash Point 51°C
Boiling Point 170°C to 172°C
Specific Gravity 0.98
Density 0.9800g/mL
Color Colorless to Yellow
Packaging Glass bottle
Infrared Spectrum Authentic
Quantity 5g
CAS Min % 98.5
CAS Max % 100.0
Assay Percent Range 99%
Linear Formula CH3C6H4OCH3
Beilstein 06, 352
Solubility Information Solubility in water: immiscible
Formula Weight 122.17
Physical Form Liquid
Percent Purity ≥98.5%
Chemical Name or Material 2-Methylanisole, 99%
Show More Show Less

GHS H Statement:
Flammable liquid and vapor.

GHS P Statement:
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

EINECSNumber : 209-426-3

TSCA : TSCA