Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Methylbenzotrifluoride, 97%

Catalog No. p-7027018
Click to view available options
Quantity:
1 g
5 g

CAS: 13630-19-8 | C8H7F3 | 160.14 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13630-19-8
Molecular Formula C8H7F3
Molecular Weight (g/mol) 160.14
MDL Number MFCD00153204
InChI Key DVFVNJHIVAPTMS-UHFFFAOYSA-N
Synonym 2-methylbenzotrifluoride, 1-methyl-2-trifluoromethyl benzene, 2-trifluoromethyl toluene, 1-methyl-2-trifluoromethyl-benzene, alpha,alpha,alpha-trifluoro-o-xylene, benzene, methyl trifluoromethyl, benzene, 1-methyl-2-trifluoromethyl, 2-methyl-1-trifluoromethyl benzene, trifluoromethyltoluene, pubchem1657
PubChem CID 2775324
IUPAC Name 1-methyl-2-(trifluoromethyl)benzene
SMILES CC1=CC=CC=C1C(F)(F)F

Specifications

Density 1.174
Boiling Point 125°C to 126°C
Refractive Index 1.435
Quantity 1 g
UN Number UN1993
Beilstein 1940501
Formula Weight 160.14
Percent Purity 97%
Chemical Name or Material 2-Methylbenzotrifluoride
Hazard Category H226-H315-H319-H335
Hazard Statement GHS H Statement
H226-H315-H319-H335
Flammable liquid and vapour.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P210-P233-P235-P240-P241-P242-P243-P261-P264b-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P312-P332+P313-P363-P370+P378q-P501c
DOTInformation Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: FLAMMABLE LIQUIDS, N.O.S.
EINECSNumber 000-000-0
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.