Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Methylbutane (Certified ACS), Fisher Chemical

Catalog No. O35514 Shop All Fisherbrand Products
Change view
Click to view available options
Quantity:
4 L
Packaging:
Glass Bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
O3551-4 4 L Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. O3551-4

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Chemical Identifiers

CAS 78-78-4
Molecular Formula C5H12
Molecular Weight (g/mol) 72.15
MDL Number MFCD00009338
InChI Key QWTDNUCVQCZILF-UHFFFAOYSA-N
Synonym isopentane, isoamylhydride, butane, 2-methyl, dimethylethylmethane, ethyldimethylmethane, 1,1,2-trimethylethane, iso-pentane, 1,1-dimethylpropane, butanes, iso-c5h12
PubChem CID 6556
ChEBI CHEBI:30362
IUPAC Name 2-methylbutane
SMILES CCC(C)C

Specifications

Alpha Vector Methylbutane
Melting Point -160°C
Color Colorless
Density 0.620g/cm³
Boiling Point 30°C
Flash Point −51°C
Assay Percent Range ≥95 %
Packaging Glass Bottle
Linear Formula (CH3)2CHCH2CH3
Quantity 4 L
UN Number 1265
Vapor Pressure 990mbar at 20°C
Formula Weight 72.15
Percent Purity ≥95%
Grade Certified
Physical Form Liquid
Chemical Name or Material 2-Methylbutane
Show More Show Less
Safety handling Solvent Safety handling Check Alert storage code Red
Hazard Statement DANGER!
Precautionary Statement Emergency Overview
Harmful by inhalation, in contact with skin and if swallowed. Keep container tightly closed. Ensure adequate ventilation. . . . . .

NFPA

Health:1
Flammability:4
Instability:0
DOTInformation Class 3: Flammable Liquid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.