Learn More
2-Methylbutane, CHROMASOLV™, for HPLC, ≥99.5%, Solstice
Shop All Solstice Research Chemicals ProductsDescription
- For HPLC and organic synthesis
- High-purity, multipurpose solvents
- High lot-to-lot consistency
Specifications
Specifications
| Absorbance | max. 0.01 at 400 nm, max. 0.01 at 240 nm, max. 0.07 at 220 nm, max. 0.30 at 210 nm, max. 1.0 at 192 nm |
| CAS Max % | 100.0000% |
| Density | 0.62 g/mL at 25 °C(lit.) |
| Boiling Point | 28°C |
| Flash Point | −51°C |
| Packaging | Glass Bottle |
| Linear Formula | CH3CH2CH(CH3)2 |
| Quantity | 1 L |
| UN Number | UN1265 |
| Beilstein | 1730723 |
| Show More |
Safety and Handling
| ShelfLife | 1260 days from date of manufacture |
| DOTInformation | Transport Hazard Class: 3; Packing Group:3: I; Proper Shipping Name: Pentanes |
| EINECSNumber | 201-142-8 |
| Recommended Storage | Room Temp |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.