Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
2-Methylcyclohexanone 96.0+%, TCI America™
Shop All Tokyo Chemical Industry (TCI) Products

Click to view available options
Quantity:
25 mL
100 mL
Specifications
Specifications
Color | Colorless |
Boiling Point | 165°C |
Quantity | 100 mL |
UN Number | 2297 |
Formula Weight | 112.17 |
Percent Purity | ≥96.0% (GC) |
Physical Form | Liquid |
Chemical Name or Material | 2-Methylcyclohexanone |
Safety and Handling
EINECSNumber | (3)-2411 |
RTECSNumber | GW1750000 |
TSCA | Yes |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction