Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2'-Methylformanilide 98.0+%, TCI America™
SDP

Catalog No. M143125G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M143125G 25 g
M14315G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M143125G Supplier TCI America Supplier No. M143125G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 94-69-9
Molecular Formula C8H9NO
Molecular Weight (g/mol) 135.17
MDL Number MFCD00014122
InChI Key ZXTLGJAARBNQGK-UHFFFAOYSA-N
Synonym 2'-methylformanilide, 2-methylformanilide, o-tolylformamide, n-2-methylphenyl formamide, formamide, n-2-methylphenyl, o-methylformanilide, o-formotoluidide, o-methyl-n-formylaniline, 2-methylphenylformamide, n-formyl-2-methylaniline
PubChem CID 7202
IUPAC Name N-(2-methylphenyl)formamide
SMILES CC1=CC=CC=C1NC=O

Specifications

Melting Point 61°C
Quantity 25 g
Formula Weight 135.17
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2′-Methylformanilide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.