Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Methylglutaric acid, 98%

Catalog No. AAA1182314
Click to view available options
Quantity:
5 g
25 g
100 g

CAS: 617-62-9 | C6H10O4 | 146.142 g/mol

2-Methylglutaric acid is used as primary and secondary intermediates in pharmaceutical and chemical research.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Methylglutaric acid is used as primary and secondary intermediates in pharmaceutical and chemical research.

Solubility
Soluble in water.

Notes
Store away from oxidizing agents. Incompatible with bases and oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 617-62-9
Molecular Formula C6H10O4
Molecular Weight (g/mol) 146.142
MDL Number MFCD00002661
InChI Key AQYCMVICBNBXNA-UHFFFAOYSA-N
Synonym 2-methylglutaric acid, alpha-methylglutaric acid, pentanedioic acid, 2-methyl, glutaric acid, 2-methyl, 2-methyl-glutaric acid, 2-methylglutarate, .alpha.-methylglutaric acid, 2-methyl glutaric acid, 2-methyl-pentanedioic acid, 2-methylpentanedioate
PubChem CID 12046
ChEBI CHEBI:68567
IUPAC Name 2-methylpentanedioic acid
SMILES CC(CCC(=O)O)C(=O)O

Specifications

Melting Point 74°C to 78°C
Boiling Point 214°C to 215°C (22 mmHg)
Quantity 25 g
Beilstein 1723219
Solubility Information Soluble in water.
Formula Weight 146.14
Percent Purity 98%
Chemical Name or Material 2-Methylglutaric acid
Hazard Category H302-H314-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P260-P264b-P270-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P501c
EINECSNumber 210-521-7
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.