Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Methylheptanoic acid, 98%

Catalog No. AAB2205014
Click to view available options
Quantity:
5 g
25 g
100 g

CAS: 1188-02-9 | C8H16O2 | 144.214 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1188-02-9
Molecular Formula C8H16O2
Molecular Weight (g/mol) 144.214
MDL Number MFCD00142944
InChI Key NKBWMBRPILTCRD-UHFFFAOYSA-N
Synonym 2-methylenanthic acid, heptanoic acid, 2-methyl, methylamylacetic acid, 2-methyl-heptanoic acid, heptanoic acid, methyl, alpha-methylheptanoic acid, 2-methyl heptanoic acid, heptane-2-carboxylic acid, fema no. 2706, heptanoic acid, 2-methyl-, 2s
PubChem CID 14475
IUPAC Name 2-methylheptanoic acid
SMILES CCCCCC(C)C(=O)O

Specifications

Density 0.906
Boiling Point 139°C to 141°C (30 mmHg)
Flash Point >110°C (230°F)
Refractive Index 1.4255
Quantity 25 g
Formula Weight 144.22
Percent Purity 98%
Chemical Name or Material 2-Methylheptanoic acid
Hazard Category H315-H318-H335
Hazard Statement GHS H Statement
H318-H335-H315
Causes serious eye damage.
May cause respiratory irritation.
Causes skin irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P310-P312-P332+P313-P362-P501c
EINECSNumber 214-704-2
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.