Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Methylhexanoic acid, 98%

Catalog No. p-7081354
Click to view available options
Quantity:
5 g
25 g

CAS: 4536-23-6 | C7H13O2 | 129.18 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 4536-23-6
Molecular Formula C7H13O2
Molecular Weight (g/mol) 129.18
MDL Number MFCD00002674
InChI Key CVKMFSAVYPAZTQ-ZCFIWIBFSA-M
Synonym 2-methylcaproic acid, hexanoic acid, 2-methyl, methylhexanoic acid, 2-methyl hexanoic acid, 2-hexanecarboxylic acid, alpha-methylcaproic acid, 2-methyl-hexanoic acid, fema no. 3191, .alpha.-methylcaproic acid, hexanoic acid, methyl
PubChem CID 20653
SMILES CCCC[C@@H](C)C([O-])=O

Specifications

Density 0.916
Boiling Point 209°C to 210°C
Flash Point 105°C (221°F)
Odor Rancid
Refractive Index 1.4205
Quantity 5 g
UN Number UN3265
Beilstein 1721227
IUPAC Name 2-methylhexanoic acid
Formula Weight 130.19
Percent Purity 98%
Chemical Name or Material 2-Methylhexanoic acid
Show More Show Less
Hazard Category H226-H314-H335
Hazard Statement GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
Precautionary Statement P210-P233-P235-P240-P241-P242-P243-P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P370+P378q-P501c
DOTInformation Hazard Class: 8; Packaging Group: II
EINECSNumber 224-883-9
RTECSNumber MO8400600
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.