Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Methylindene, Thermo Scientific Chemicals

Catalog No. p-7214780
Click to view available options
Quantity:
1 g
5 g

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2177-47-1
Molecular Formula C10H10
Molecular Weight (g/mol) 130.19
MDL Number MFCD00274253
InChI Key YSAXEHWHSLANOM-UHFFFAOYSA-N
IUPAC Name 2-methyl-1H-indene
SMILES CC1=CC2=CC=CC=C2C1

Specifications

Density 0.9710 g/mL
Flash Point 76°C
Quantity 1 g
Specific Gravity 0.971
Formula Weight 130.19
Chemical Name or Material 2-Methylindene

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.