Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Methylindole-3-carboxaldehyde 98.0+%, TCI America™
SDP

Catalog No. M148525G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M148525G 25 g
M14855G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M148525G Supplier TCI America Supplier No. M148525G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 5416-80-8
Molecular Formula C10H9NO
Molecular Weight (g/mol) 159.188
MDL Number MFCD00012077
InChI Key CYZIVXOEJNAIBS-UHFFFAOYSA-N
Synonym 2-methylindole-3-carboxaldehyde, 3-formyl-2-methylindole, 2-methyl-3-formylindole, 1h-indole-3-carboxaldehyde, 2-methyl, 2-methylindole-3-carbaldehyde, indole-3-carboxaldehyde, 2-methyl, pubchem7437, zlchem 1339, acmc-1apkw, ksc271e8j
PubChem CID 73166
IUPAC Name 2-methyl-1H-indole-3-carbaldehyde
SMILES CC1=C(C2=CC=CC=C2N1)C=O

Specifications

Melting Point 203°C
Quantity 25 g
Formula Weight 159.19
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2-Methylindole-3-carboxaldehyde
EINECSNumber (5)-0136
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.