Learn More
2-Methylindole, 98+%
Description
2-Methylindole is used as a reactant for regioselective synthesis of oxopyrrolidine analogs via iodine-catalyzed Markovnikov addition reaction, Friedel-Crafts alkylation reactions, preparation of tryptophan dioxygenase inhibitors pyridyl-ethenyl-indoles as potential anticancer immunomodulators, Michael addition reactions and in synthesis of cyclooxygenase-1 (COX-1)/cyclooxygenase-2 (COX-2) inhibitors.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
2-Methylindole is used as a reactant for regioselective synthesis of oxopyrrolidine analogs via iodine-catalyzed Markovnikov addition reaction, Friedel-Crafts alkylation reactions, preparation of tryptophan dioxygenase inhibitors pyridyl-ethenyl-indoles as potential anticancer immunomodulators, Michael addition reactions and in synthesis of cyclooxygenase-1 (COX-1)/cyclooxygenase-2 (COX-2) inhibitors.
Solubility
Insoluble in water.
Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents. It is sensitive to light.

Specifications
Specifications
Melting Point | 56°C to 60°C |
Density | 1.07 |
Boiling Point | 272°C to 273°C |
Flash Point | 141°C (285°F) |
Quantity | 250 g |
Beilstein | 109781 |
Sensitivity | Air and light sensitive |
Solubility Information | Insoluble in water. |
Formula Weight | 131.18 |
Percent Purity | ≥98% |
Show More |
Safety and Handling
Hazard Category | H302-H315-H319-H335 |
Hazard Statement | GHS H Statement H301 Toxic if swallowed. |
Precautionary Statement | P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c |
DOTInformation | Hazard Class: 9; Packaging Group: III |
EINECSNumber | 202-398-3 |
RTECSNumber | NM0345000 |
TSCA | Yes |
Recommended Storage | Ambient temperatures; Store under Argon |
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.