Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Methylindole, 98%

Catalog No. AC127025000
Click to view available options
Quantity:
100 g
500 g
Packaging:
Glass bottle

CAS: 95-20-5 | C9H9N | 131.18 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 95-20-5
Molecular Formula C9H9N
Molecular Weight (g/mol) 131.18
MDL Number MFCD00005616
InChI Key BHNHHSOHWZKFOX-UHFFFAOYSA-N
Synonym 2-methylindole, 1h-indole, 2-methyl, indole, 2-methyl, 2-methyl indole, unii-i7cn58827i, 2-methylindol, methylketole, methyl indole, 2-methyl-indole, pubchem7504
PubChem CID 7224
ChEBI CHEBI:49402
IUPAC Name 2-methyl-1H-indole
SMILES CC1=CC2=CC=CC=C2N1

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point 56.0°C to 60.0°C
Color Brown or Purple-Red to Yellow
Boiling Point 273.0°C
Flash Point 141°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Quantity 500 g
Beilstein 20, 311
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in ethanol,ethyl ether and acetone
Formula Weight 131.18
Percent Purity 98%
Physical Form Crystalline Powder, Crystals or Flakes
Chemical Name or Material 2-Methylindole
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
May cause respiratory irritation.
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
Precautionary Statement GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 202-398-3

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.