Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-(Methylmercapto)benzimidazole 99%, Thermo Scientific™

Catalog No. AC127120100
Click to view available options
:
10g

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Specifications

CAS 7152-24-1
CAS Min % 98.5
CAS Max % 100.0
Infrared Spectrum Authentic
Molecular Formula C8H8N2S
Quantity 10g
MDL Number MFCD00005594
Synonym 2-methylthio benzimidazole, 1h-benzimidazole, 2-methylthio, 2-methylmercaptobenzimidazole, 2-methylthio-1h-benzo d imidazole, 2-methylthiobenzimidazole, 2-methylsulfanyl-1h-benzimidazole, s-methyl-2-mercaptobenzimidazole, benzimidazole, 2-methylthio
InChI Key OCKJFOHZLXIAAT-UHFFFAOYSA-N
SMILES CSC1=NC2=CC=CC=C2N1
IUPAC Name 2-methylsulfanyl-1H-benzimidazole
Molecular Weight (g/mol) 164.226
PubChem CID 23539
Formula Weight 164.22
Percent Purity ≥98.5%
Chemical Name or Material 2-(Methylmercapto)benzimidazole, 99%
Show More Show Less
EINECSNumber 230-494-5
RTECSNumber DD9480000
TSCA TSCA
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.