Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

6,9-Diamino-2-ethoxyacridine lactate monohydrate, 95%, Thermo Scientific Chemicals

Catalog No. AC406380100
Click to view available options
Quantity:
5 g
10 g
100 g
Packaging:
Glass bottle

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 6402-23-9, 1837-57-6
Molecular Formula C3H6O3·H2O
Molecular Weight (g/mol) 361.39
MDL Number MFCD00149646
InChI Key NYEPHMYJRNWPLA-UHFFFAOYSA-N
Synonym ethacridine lactate monohydrate, ethodin, ethacridine lactate hydrate, ethacridine lactate salt, dsstox_cid_26274, dsstox_rid_81501, dsstox_gsid_46274, 7-ethoxyacridine-3,9-diamine; 2-hydroxypropanoic acid; hydrate, rivanol monohydrate
PubChem CID 165457
IUPAC Name 7-ethoxyacridine-3,9-diamine;2-hydroxypropanoic acid;hydrate
SMILES CCOC1=CC2=C(C3=C(C=C(C=C3)N)N=C2C=C1)N.CC(C(=O)O)O.O

Specifications

CAS Min % 94.0
CAS Max % 100.0
Melting Point 243°C to 245°C
Color Yellow
Infrared Spectrum Authentic
Assay Percent Range 95%
Packaging Glass bottle
Quantity 10 g
Merck Index 15, 3771
Solubility Information Solubility in water: .. Other solubilities: slowly soluble in 15 parts water,soluble in 9 parts boiling water,soluble in 110 parts alcohol (22°C),soluble in 100 parts alcohol (20°C)
Formula Weight 361.39
Percent Purity 95%
Physical Form Powder
Chemical Name or Material 6, 9-Diamino-2-ethoxyacridine lactate monohydrate
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
Harmful if swallowed.
Harmful if inhaled.
Causes skin irritation.
May cause respiratory irritation.
Harmful in contact with skin.
Causes serious eye irritation.
Precautionary Statement GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.