Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Methylpyridine-4-boronic acid pinacol ester, 95%, Thermo Scientific Chemicals

Catalog No. AC448722500
Click to view available options
Quantity:
250 mg
1 g

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 660867-80-1
Molecular Formula C12H18BNO2
Molecular Weight (g/mol) 219.09
MDL Number MFCD08061590
InChI Key MBTULFIFECUURA-UHFFFAOYSA-N
Synonym 2-methylpyridine-4-boronic acid pinacol ester, 2-methyl-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl pyridine, 2-methyl-4-tetramethyl-1,3,2-dioxaborolan-2-yl pyridine, pyridine, 2-methyl-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl, 2-methyl-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl-pyridine, 2-methylpyridine-4-boronic acid pinacol, 2-methyl-4-pyridineboronic acid pinacol ester, 2-methylpyridin-4-ylboronic acid pinacol ester, 2-methylpyridin-4-yl boronic acid pinacol ester, 2-methylpyridine-4-yl boronic acid pinacol ester
PubChem CID 16414274
IUPAC Name 2-methyl-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridine
SMILES CC1=CC(=CC=N1)B1OC(C)(C)C(C)(C)O1

Specifications

Melting Point 66.0°C to 71.0°C
Assay Percent Range 94% min. (GC)
Packaging Glass bottle
Quantity 250 mg
Solubility Information Solubility in water: insoluble
Formula Weight 219.09
Percent Purity 95%
Chemical Name or Material 2-Methylpyridine-4-boronic acid pinacol ester
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Harmful if swallowed.
Precautionary Statement GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.