Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Methylpyrimidine-5-carboxylic acid, 97%

Catalog No. AAH64375MD
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH64375MD 250 mg
AAH6437503 1 g
AAH6437506 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAH64375MD Supplier Thermo Scientific Chemicals Supplier No. H64375MD

CAS: 5194-32-1 | C6H6N2O2 | 138.126 g/mol

It is employed in the synthesis, spectral characterization and anticancer activity of new 2,3,6- substituted quinazolin-4(3h)-one derivatives.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is employed in the synthesis, spectral characterization and anticancer activity of new 2,3,6- substituted quinazolin-4(3h)-one derivatives.

Solubility
Slightly soluble in water.

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition. Protect from heat. Store in freezer at -20°C.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 5194-32-1
Molecular Formula C6H6N2O2
Molecular Weight (g/mol) 138.126
MDL Number MFCD08235210
InChI Key NMGIXZFBQPETOK-UHFFFAOYSA-N
Synonym 2-methyl-5-pyrimidinecarboxylic acid, 2-methylpyrimidine-5-carboxylicacid, 5-pyrimidinecarboxylic acid, 2-methyl, 2-methyl-5-pyrimidinecarboxylicacid, pubchem13290, acmc-209kwu, ksc269i7t, 2-methyl-pyrimidine-5-carboxylic acid, 5-pyrimidinecarboxylicacid, 2-methyl, 2-methyl pyrimidine-5-carboxylic acid
PubChem CID 13670875
IUPAC Name 2-methylpyrimidine-5-carboxylic acid
SMILES CC1=NC=C(C=N1)C(=O)O

Specifications

Melting Point 203°C to 205°C
Quantity 250 mg
Beilstein 508773
Solubility Information Slightly soluble in water.
Formula Weight 138.13
Percent Purity 97%
Physical Form Solid
Chemical Name or Material 2-Methylpyrimidine-5-carboxylic acid
Hazard Category H302-H315-H319-H335
Hazard Statement GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
TSCA No
storageNote1 -30° to -10°C
Recommended Storage Store at -20°C

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.