Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Methylpyrrolidine, 98%

Catalog No. p-2690117
Click to view available options
Quantity:
2 g
10 g
Packaging:
Glass bottle

CAS: 765-38-8 | C5H11N | 85.15 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 765-38-8
Molecular Formula C5H11N
Molecular Weight (g/mol) 85.15
MDL Number MFCD00014491
InChI Key RGHPCLZJAFCTIK-UHFFFAOYSA-N
PubChem CID 13003
IUPAC Name 2-methylpyrrolidine
SMILES CC1CCCN1

Specifications

CAS Min % 97.5
CAS Max % 100.0
Color Colorless to Yellow
Density 0.8800g/mL
Boiling Point 98°C to 100°C
Flash Point 2°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Refractive Index 1.4350 to 1.437
Quantity 2 g
Specific Gravity 0.88
Synonym pyrrolidine, 2-methyl, 2-methyl-pyrrolidine, 2-methylpyrolidine, 2-methyl-pyrolidine, 2-methyl pyrrolidine, pubchem9410, pyrrolidine,2-methyl, acmc-209mch, acmc-1aspi, rac-2-methyl-pyrrolidine
Solubility Information Solubility in water: soluble in water. Other solubilities: soluble in common organic solvents
Formula Weight 85.15
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material 2-Methylpyrrolidine, 98%
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement:
Causes serious eye damage.
Causes severe skin burns and eye damage.
Highly flammable liquid and vapor.
Precautionary Statement GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Ground/Bond container and receiving equipment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 212-144-3

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.