Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Methyltetrahydrofuran-3-thiol, 97%, mixed isomers, ACROS Organics™

Click to view available options
Quantity:
1g
5g
25g
Packaging:
Glass bottle
57124-87-5
C5H10OS
118.2
MFCD01632596
DBPHPBLAKVZXOY-UHFFFAOYSA-N
2-methyl-3-tetrahydrofuranthiol,2-methyltetrahydrofuran-3-thiol,tetrahydro-2-methyl-3-furanthiol,3-furanthiol, tetrahydro-2-methyl,2-methyl-3-mercaptotetrahydrofuran,pentitol, 1,4-anhydro-2,5-dideoxy-3-thio,3-mercapto-2-methyltetrahydrofuran,pubchem9972,ksc201q4p,2-methyl-3-tetrahydrofuran thiol
62128
2-methyloxolane-3-thiol
CC1C(CCO1)S
57124-87-5
C5H10OS
118.2
MFCD01632596
DBPHPBLAKVZXOY-UHFFFAOYSA-N
2-methyl-3-tetrahydrofuranthiol,2-methyltetrahydrofuran-3-thiol,tetrahydro-2-methyl-3-furanthiol,3-furanthiol, tetrahydro-2-methyl,2-methyl-3-mercaptotetrahydrofuran,pentitol, 1,4-anhydro-2,5-dideoxy-3-thio,3-mercapto-2-methyltetrahydrofuran,pubchem9972,ksc201q4p,2-methyl-3-tetrahydrofuran thiol
62128
2-methyloxolane-3-thiol
CC1C(CCO1)S
Specific Gravity 1.04
Packaging Glass bottle
Color Colorless to Yellow
Flash Point 40°C
Boiling Point 160.0°C to 180.0°C
Quantity 25g
Infrared Spectrum Authentic
Density 1.0400g/mL
Refractive Index 1.4730 to 1.4910
CAS Min % 96.0
CAS Max % 100.0
Assay Percent Range 97%
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in most organic solvents
Formula Weight 118.2
Physical Form Liquid
Percent Purity 97%
Chemical Name or Material 2-Methyltetrahydrofuran-3-thiol, Mixture of isomers
Show More Show Less

GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
Flammable liquid and vapor.
May cause respiratory irritation.

GHS P Statement
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present a

GHS Signal Word: Warning

EINECSNumber : 260-572-4