Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Methyltetrahydrofuran-3-thiol, 97%, mixed isomers, Thermo Scientific™

Catalog No. p-3779219
Click to view available options
Quantity:
1 g
25 g
Packaging:
Glass bottle

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 57124-87-5
Molecular Formula C5H10OS
Molecular Weight (g/mol) 118.2
MDL Number MFCD01632596
InChI Key DBPHPBLAKVZXOY-UHFFFAOYSA-N
Synonym 2-methyl-3-tetrahydrofuranthiol, 2-methyltetrahydrofuran-3-thiol, tetrahydro-2-methyl-3-furanthiol, 3-furanthiol, tetrahydro-2-methyl, 2-methyl-3-mercaptotetrahydrofuran, pentitol, 1,4-anhydro-2,5-dideoxy-3-thio, 3-mercapto-2-methyltetrahydrofuran, pubchem9972, ksc201q4p, 2-methyl-3-tetrahydrofuran thiol
PubChem CID 62128
IUPAC Name 2-methyloxolane-3-thiol
SMILES CC1C(CCO1)S

Specifications

Color Colorless to Yellow
CAS Min % 96.0
CAS Max % 100.0
Density 1.0400g/mL
Boiling Point 160.0°C to 180.0°C
Quantity 25 g
Infrared Spectrum Authentic
Packaging Glass bottle
Physical Form Liquid
Refractive Index 1.4730 to 1.4910
Assay Percent Range 97%
Specific Gravity 1.04
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in most organic solvents
Formula Weight 118.2
Percent Purity 97%
Chemical Name or Material 2-Methyltetrahydrofuran-3-thiol, Mixture of isomers
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
Flammable liquid and vapor.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present a
EINECSNumber 260-572-4

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.