Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Methyltetrahydrofuran, ≥99.5% (GC), MilliporeSigma™ Supelco™

Catalog No. p-7187791
Click to view available options
Quantity:
5 mL
10 mL

2-Methyltetrahydrofuran is a sustainable green alternative and offers both economical and environmentally friendly advantages over tetrahydrofuran and dichloromethane in a variety of organometallic reactions and biphasic reactions.

Environmental
TRUSTED_SUSTAINABILITY

Specifications

CAS 96-47-9
Density 0.86 g/mL (at 25°C)
Flash Point -10°C
Physical Form Neat
Linear Formula C5H10 O
MDL Number MFCD00005367
Boiling Point 78°C to 80°C
Synonym 2-MeTHF; Tetrahydro-2-methylfuran; Tetrahydrosilvan
Type Analytical Standard
Quantity 5 mL
Formula Weight 86.13
Percent Purity ≥99.5% (GC)
Grade Analytical Standard
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.