Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
2-Methyltetrahydrofuran, Spectrum™ Chemical
Shop All Spectrum Chemical Manufacturing Corporation Products

Click to view available options
Quantity:
100 mL
1 L
Packaging:
Amber Glass Bottle
Description
Spectrum™ Chemical 2-Methyltetrahydrofuran
Specifications
Specifications
CAS Max % | 100% |
Quantity | 100 mL |
Packaging | Amber Glass Bottle |
UN Number | UN2536 |
Grade | Reagent |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction